Assay Details
Target Gene Details
Entrez Gene ID: | 16440 |
Gene Name: | inositol 1,4,5-triphosphate receptor 3 |
Gene Aliases: |
Ip3r3, Itpr-3 |
Location: |
Chr.17:27057260-27122223 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itpr3 | NM_080553.3 | 1 | 311 | NP_542120.2 |
| AB182289.1 | 1 | 79 | BAD90683.1 | |
| AK134443.1 | 1 | 316 | ||
| BB866373.1 | 1 | 311 |
More Information
Set Membership: |
|

Genomic Map