Assay Details
Target Gene Details
Entrez Gene ID: | 19326 |
Gene Name: | RAB11B, member RAS oncogene family |
Gene Aliases: |
A730055L17Rik |
Location: |
Chr.17:33742484-33760486 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab11b | NM_008997.3 | 1 | 128 | NP_033023.1 |
| AK134125.1 | BAE22024.1 | |||
| AK146452.1 | BAE27183.1 | |||
| AK146460.1 | BAE27188.1 | |||
| AK146623.1 | BAE27312.1 | |||
| AK147077.1 | BAE27658.1 | |||
| AK148022.1 | BAE28293.1 | |||
| BC054753.1 | AAH54753.1 | |||
| BC157916.1 | ||||
| BU705914.1 | 1 | 128 | ||
| BY275360.1 | 1 | 123 | ||
| L26528.1 | AAC42093.1 |

Genomic Map