Assay Details
Target Gene Details
Entrez Gene ID: | 19326 |
Gene Name: | RAB11B, member RAS oncogene family |
Gene Aliases: |
A730055L17Rik |
Location: |
Chr.17:33742484-33760486 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab11b | NM_008997.3 | 5 | 3414 | NP_033023.1 |
| AK036482.1 | 1 | 624 | ||
| AK043096.1 | 1 | 780 |

Genomic Map