Assay Details
Target Gene Details
Entrez Gene ID: | 22154 |
Gene Name: | tubulin, beta 5 class I |
Gene Aliases: |
AA408537, AI596182, B130022C14Rik, M(beta)5 |
Location: |
Chr.17:35833920-35838301 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb5 | NM_011655.5 | 1 | 158 | NP_035785.1 |
| AK010960.1 | 1 | 90 | BAB27292.1 | |
| AK011263.1 | 1 | 86 | BAB27504.1 | |
| AK011765.1 | 1 | 90 | ||
| AK012350.1 | 1 | 90 | ||
| AK017806.1 | 1 | 90 | ||
| AK051164.1 | 1 | 90 | BAC34541.1 | |
| AK051393.1 | 1 | 92 | BAC34623.1 | |
| AK076196.1 | 1 | 85 | ||
| AK083327.1 | 1 | 90 | BAC38866.1 | |
| AK146567.1 | 1 | 90 | BAE27265.1 | |
| AK148978.1 | 1 | 85 | BAE28709.1 | |
| AK151215.1 | 1 | 90 | BAE30210.1 | |
| AK151670.1 | 1 | 90 | BAE30596.1 | |
| AK160225.1 | 1 | 90 | BAE35700.1 | |
| AK161575.1 | 1 | 90 | BAE36471.1 | |
| AK165249.1 | 1 | 88 | BAE38103.1 | |
| AK167691.1 | 1 | 90 | BAE39738.1 | |
| AK167779.1 | 1 | 90 | BAE39811.1 | |
| AK167808.1 | 1 | 85 | BAE39835.1 | |
| AK167926.1 | 1 | 90 | BAE39931.1 | |
| AK168271.1 | 1 | 85 | BAE40217.1 | |
| AK169123.1 | 1 | 85 | BAE40903.1 | |
| AK169210.1 | 1 | 89 | BAE40982.1 | |
| AK169295.1 | 1 | 90 | BAE41051.1 | |
| AK169606.1 | 1 | 91 | ||
| AK207156.1 | ||||
| AK212131.1 | ||||
| AK213420.1 | ||||
| AK215963.1 | ||||
| AK216210.1 | ||||
| BC003825.1 | 1 | 64 | AAH03825.1 | |
| BY111488.1 | 1 | 158 | ||
| X04663.1 | CAA28369.1 |
More Information
Set Membership: |
|

Genomic Map