Assay Details
Target Gene Details
Entrez Gene ID: | 66306 |
Gene Name: | family with sequence similarity 53, member C |
Gene Aliases: |
2810012G03Rik, Gm21179 |
Location: |
Chr.18:34758830-34773760 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam53c | NM_175104.4 | NP_780313.1 | ||
| BC057111.1 | AAH57111.1 |
More Information
Set Membership: |
|

Genomic Map