Assay Details
Target Gene Details
Entrez Gene ID: | 68591 |
Gene Name: | molybdenum cofactor sulfurase |
Gene Aliases: |
1110018O12Rik, HMCS, MCS, MOS |
Location: |
Chr.18:24653176-24701577 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mocos | NM_026779.1 | 8 | 1854 | NP_081055.1 |
| AK076385.1 | 8 | 1668 | ||
| BC113180.1 | 8 | 1854 | ||
| BC113786.1 | 8 | 1854 |

Genomic Map