Assay Details
Target Gene Details
Entrez Gene ID: | 210741 |
Gene Name: | potassium channel, subfamily K, member 12 |
Gene Aliases: |
mntk1 |
Location: |
Chr.17:87745821-87797994 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk12 | NM_199251.1 | 1 | 155 | NP_954859.1 |
| AB052939.1 | 1 | 155 | BAD06001.1 |
More Information
Set Membership: |
|

Genomic Map