Assay Details
Target Gene Details
Entrez Gene ID: | 210741 |
Gene Name: | potassium channel, subfamily K, member 12 |
Gene Aliases: |
mntk1 |
Location: |
Chr.17:87745821-87797994 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk12 | NM_199251.1 | 2 | 1104 | NP_954859.1 |
| AB052939.1 | 2 | 1104 | BAD06001.1 | |
| BC138133.1 | 2 | 896 | ||
| BC138134.1 | 2 | 896 |
More Information
Set Membership: |
|

Genomic Map