Assay Details
Target Gene Details
Entrez Gene ID: | 19765 |
Gene Name: | ralA binding protein 1 |
Gene Aliases: |
RLIP76, Rip1 |
Location: |
Chr.17:65848415-65885755 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ralbp1 | NM_001198949.1 | 2 | 392 | NP_001185878.1 |
| NM_009067.5 | 2 | 716 | NP_033093.2 | |
| AK019116.1 | 2 | 271 | BAB31552.1 | |
| AK133613.1 | 2 | 275 | BAE21750.1 | |
| AK136116.1 | 2 | 481 | BAE22829.1 | |
| AK138197.1 | 2 | 760 | BAE23576.1 | |
| AK145387.1 | 2 | 256 | BAE26405.1 | |
| AK159404.1 | 2 | 269 | BAE35056.1 | |
| BC028516.1 | 2 | 264 | ||
| BC067073.1 | 1 | 151 | AAH67073.1 | |
| BC075732.1 | 2 | 341 | AAH75732.1 | |
| BC076636.1 | 2 | 308 | AAH76636.1 | |
| BC138546.1 | 1 | 157 | ||
| BC145320.1 | 1 | 157 | ||
| BC145815.1 | 1 | 157 | ||
| CJ146118.1 | 2 | 393 | ||
| X80937.1 | 2 | 293 | CAA56912.1 |
More Information
Set Membership: |
|

Genomic Map