Assay Details
Target Gene Details
Entrez Gene ID: | 83672 |
Gene Name: | synaptotagmin-like 3 |
Gene Aliases: |
A430092N21, Slp3, Slp3-a, Slp3-b, mFLJ00307 |
Location: |
Chr.17:6658233-6738070 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sytl3 | NM_031395.2 | 15 | 2042 | NP_113572.1 |
| NM_183368.2 | 9 | 1238 | NP_899224.2 | |
| NM_183370.2 | 13 | 1456 | NP_899226.2 | |
| AB057758.1 | 16 | 2042 | BAB41086.1 | |
| AB057764.1 | 17 | 2186 | BAB41092.1 | |
| AB057765.1 | 15 | 1963 | BAB41093.1 | |
| AK131174.1 | 14 | 6797 | BAD21424.1 | |
| BB031741.2 | 4 | 408 | ||
| BC022608.1 | 14 | 1665 | AAH22608.1 | |
| BC106102.1 | 13 | 1456 | AAI06103.1 | |
| BC118936.2 | 16 | 1867 |

Genomic Map