Assay Details
Target Gene Details
Entrez Gene ID: | 224826 |
Gene Name: | ubiquitin protein ligase E3 component n-recognin 2 |
Gene Aliases: |
9930021A08Rik, AI462103, AW540746, E130209G04Rik, mKIAA0349 |
Location: |
Chr.17:46928291-47010775 on Build GRCm38 |
Assay Gene Location: | Within Exon 48 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubr2 | NM_001177374.1 | 47 | 5804 | NP_001170845.1 |
| NM_146078.3 | 47 | 5804 | NP_666190.2 | |
| AK078002.1 | 15 | 1711 | ||
| AK147445.1 | 47 | 5793 | BAE27917.1 | |
| BC075642.1 | 48 | 5618 | AAH75642.1 |
More Information
Set Membership: |
|

Genomic Map