Assay Details
Target Gene Details
Entrez Gene ID: | 338360 |
Gene Name: | RIKEN cDNA B430212C06 gene |
Gene Aliases: |
- |
Location: |
Chr.18:67321209-67344103 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B430212C06Rik | NR_033214.1 | |||
| AK087569.1 |
Target Gene Details
Entrez Gene ID: | 12683 |
Gene Name: | cell death-inducing DNA fragmentation factor, alpha subunit-like effector A |
Gene Aliases: |
AW212747 |
Location: |
Chr.18:67343564-67367794 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cidea | NM_007702.2 | 1 | 186 | NP_031728.2 |
| AF041376.1 | 1 | 160 | AAC34985.1 | |
| BC096649.1 | AAH96649.1 | |||
| BQ554477.1 | 1 | 195 |
More Information
Set Membership: |
|

Genomic Map