Assay Details
Target Gene Details
Entrez Gene ID: | 20440 |
Gene Name: | beta galactoside alpha 2,6 sialyltransferase 1 |
Gene Aliases: |
AW742324, Siat1, St6Gal-I, St6gal, St6galI |
Location: |
Chr.16:23224728-23360350 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St6gal1 | NM_145933.4 | 1 | 177 | NP_666045.1 |
| AF153680.1 | 1 | 63 | ||
| AK084124.1 | BAC39120.1 | |||
| BC092222.1 | 1 | 155 | AAH92222.1 | |
| BC096026.1 | 1 | 75 | AAH96026.1 | |
| CF898349.1 | 1 | 177 |
More Information
Set Membership: |
|

Genomic Map