Assay Details
Target Gene Details
Entrez Gene ID: | 20440 |
Gene Name: | beta galactoside alpha 2,6 sialyltransferase 1 |
Gene Aliases: |
AW742324, Siat1, St6Gal-I, St6gal, St6galI |
Location: |
Chr.16:23224728-23360350 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St6gal1 | NM_001252505.1 | 8 | 2814 | NP_001239434.1 |
| NM_001252506.1 | 7 | 2681 | NP_001239435.1 | |
| NM_145933.4 | 8 | 3145 | NP_666045.1 | |
| AK034768.1 | 2 | 1491 | BAC28828.1 | |
| AK084124.1 | 8 | 2980 | BAC39120.1 | |
| BC027833.1 | 5 | 2342 | AAH27833.1 | |
| BC092222.1 | 8 | 3123 | AAH92222.1 | |
| BC096026.1 | 8 | 2793 | AAH96026.1 |
More Information
Set Membership: |
|

Genomic Map