Assay Details
Target Gene Details
Entrez Gene ID: | 68146 |
Gene Name: | ADP-ribosylation factor-like 13B |
Gene Aliases: |
A530097K21Rik, A930014M17Rik, Arl2l1, C530009C10Rik, hnn |
Location: |
Chr.16:62793678-62847055 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arl13b | NM_026577.3 | NP_080853.3 | ||
| AK016794.1 | BAB30434.1 | |||
| AK020859.1 | BAB32230.1 | |||
| AK021270.1 | ||||
| BC082574.1 | AAH82574.1 |
Target Gene Details
Entrez Gene ID: | 68159 |
Gene Name: | syntaxin 19 |
Gene Aliases: |
A030009B12Rik |
Location: |
Chr.16:62814676-62822722 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stx19 | NM_026588.1 | 2 | 433 | NP_080864.1 |
| AK009770.1 | 1 | 134 | BAB26492.1 | |
| AK020701.1 | 2 | 384 | BAB32182.3 | |
| BC023414.1 | 2 | 433 | AAH23414.1 |
More Information
Set Membership: |
|

Genomic Map