Assay Details
Target Gene Details
Entrez Gene ID: | 22350 |
Gene Name: | ezrin |
Gene Aliases: |
AW146364, R75297, Vil2, p81 |
Location: |
Chr.17:6738131-6782780 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ezr | NM_009510.2 | 13 | 2850 | NP_033536.2 |
| AK004588.2 | 13 | 2825 | ||
| AK035271.1 | 9 | 2218 | BAC29009.1 | |
| AK145707.1 | 13 | 2851 | BAE26602.1 | |
| AK159717.1 | 13 | 2828 | BAE35312.1 | |
| AK159937.1 | 14 | 2802 | ||
| AK166191.1 | 13 | 2826 | BAE38621.1 | |
| AK168410.1 | 13 | 2829 | ||
| AK168840.1 | 13 | 2826 | BAE40663.1 | |
| AK170610.1 | 13 | 2826 | BAE41910.1 | |
| BC048181.1 | 13 | 2813 | AAH48181.2 |
More Information
Set Membership: |
|

Genomic Map