Assay Details
Target Gene Details
Entrez Gene ID: | 15531 |
Gene Name: | N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 1 |
Gene Aliases: |
1200015G06Rik, Hsst, NDST-1, b2b2230Clo |
Location: |
Chr.18:60684494-60748892 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndst1 | NM_008306.4 | 14 | 4396 | NP_032332.2 |
| AK147282.1 | 15 | 4789 | BAE27818.1 | |
| BC066098.1 | 14 | 4396 | AAH66098.1 | |
| BC079561.1 | 14 | 4396 | AAH79561.1 |
More Information
Set Membership: |
|

Genomic Map