Assay Details
Target Gene Details
Entrez Gene ID: | 207227 |
Gene Name: | syntaxin binding protein 5-like |
Gene Aliases: |
9430094L02, A830015P08Rik, LLGL4, T2dm1, t2md1, tomosyn-2 |
Location: |
Chr.16:37107307-37384988 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stxbp5l | NM_001114611.1 | 26 | 7623 | NP_001108083.1 |
| NM_001114612.1 | 27 | 7794 | NP_001108084.1 | |
| NM_001114613.1 | 27 | 7695 | NP_001108085.1 | |
| NM_172440.3 | 28 | 7866 | NP_766028.2 |
More Information
Set Membership: |
|

Genomic Map