Assay Details
Target Gene Details
Entrez Gene ID: | 12575 |
Gene Name: | cyclin-dependent kinase inhibitor 1A (P21) |
Gene Aliases: |
CAP20, CDKI, CIP1, Cdkn1, P21, SDI1, Waf1, mda6, p21Cip1, p21WAF |
Location: |
Chr.17:29090986-29100722 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdkn1a | NM_001111099.1 | 2 | 481 | NP_001104569.1 |
| NM_007669.4 | 2 | 455 | NP_031695.1 | |
| AB017817.1 | 1 | 354 | BAA82341.1 | |
| AB017818.1 | 1 | 354 | BAA82342.1 | |
| AK007630.1 | 2 | 453 | BAB25148.1 | |
| AK089093.1 | 2 | 453 | BAC40747.1 | |
| AK139306.1 | 2 | 456 | BAE23949.1 | |
| AK149910.1 | 2 | 456 | BAE29160.1 | |
| AK170467.1 | 2 | 456 | BAE41815.1 | |
| AK194916.1 | ||||
| BC002043.1 | 2 | 408 | AAH02043.1 | |
| CT010218.1 | 1 | 354 | CAJ18426.1 | |
| U09507.1 | 1 | 555 | AAB60456.1 | |
| U24173.1 | 2 | 446 | AAC52220.1 |
More Information
Set Membership: |
|

Genomic Map