Assay Details
Target Gene Details
Entrez Gene ID: | 67544 |
Gene Name: | family with sequence similarity 120, member B |
Gene Aliases: |
4932442K08Rik, AW049283, AW413338, CCPG, Pgcc1, mKIAA1838 |
Location: |
Chr.17:15396157-15433581 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam120b | NM_024203.3 | 1 | 266 | NP_077165.2 |
| NR_033586.1 | 1 | 310 | ||
| AK016568.1 | 1 | 235 | ||
| AK028911.1 | 1 | 196 | BAC26191.1 | |
| AK051767.1 | 1 | 248 | ||
| AK076185.1 | 1 | 234 | BAC36239.1 | |
| AK078215.1 | 1 | 261 | BAC37177.1 | |
| AK087438.1 | 1 | 212 | ||
| AK132211.1 | 1 | 244 | BAE21035.1 | |
| AK161912.1 | 1 | 258 | ||
| AK220425.1 | 1 | 228 | BAD90470.1 | |
| BC085163.1 | 1 | 264 | AAH85163.1 | |
| BY073650.1 | 1 | 267 | ||
| CB518733.1 | 1 | 310 | ||
| DQ873694.1 | 1 | 247 |

Genomic Map