Assay Details
Target Gene Details
Entrez Gene ID: | 67544 |
Gene Name: | family with sequence similarity 120, member B |
Gene Aliases: |
4932442K08Rik, AW049283, AW413338, CCPG, Pgcc1, mKIAA1838 |
Location: |
Chr.17:15396157-15433581 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam120b | NM_024203.3 | 11 | 4213 | NP_077165.2 |
| NR_033586.1 | 11 | 5525 | ||
| BC085163.1 | 11 | 5478 | AAH85163.1 |

Genomic Map