Assay Details
Target Gene Details
Entrez Gene ID: | 67544 |
Gene Name: | family with sequence similarity 120, member B |
Gene Aliases: |
4932442K08Rik, AW049283, AW413338, CCPG, Pgcc1, mKIAA1838 |
Location: |
Chr.17:15396157-15433581 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam120b | NM_024203.3 | 2 | 1241 | NP_077165.2 |
| NR_033586.1 | 2 | 1285 | ||
| AK016568.1 | 2 | 1209 | ||
| AK028911.1 | 2 | 1171 | BAC26191.1 | |
| AK050319.1 | 1 | 956 | BAC34186.1 | |
| AK076185.1 | 2 | 1207 | BAC36239.1 | |
| AK087438.1 | 2 | 1187 | ||
| AK132211.1 | 2 | 1219 | BAE21035.1 | |
| AK220425.1 | 2 | 1203 | BAD90470.1 | |
| BC003200.1 | 1 | 147 | AAH03200.1 | |
| BC085163.1 | 2 | 1239 | AAH85163.1 | |
| DQ873694.1 | 2 | 1222 |

Genomic Map