Assay Details
Target Gene Details
Entrez Gene ID: | 72775 |
Gene Name: | Fanconi anemia, complementation group E |
Gene Aliases: |
2810451D06Rik, AI415634, AW209126 |
Location: |
Chr.17:28313530-28326574 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fance | NM_001163819.1 | 2 | 644 | NP_001157291.1 |
| NM_001163820.1 | 2 | 644 | NP_001157292.1 | |
| NR_028296.1 | 2 | 644 | ||
| NR_028297.1 | 2 | 644 | ||
| AK013325.1 | 2 | 608 | ||
| AK135017.1 | 3 | 548 | BAE22385.1 | |
| AK172283.1 | 2 | 628 | BAE42924.1 | |
| AK182639.1 | ||||
| AK213002.1 | ||||
| BC016538.1 | 2 | 602 | AAH16538.1 | |
| BY740269.1 | 2 | 628 |

Genomic Map