Assay Details
Target Gene Details
Entrez Gene ID: | 214505 |
Gene Name: | N-acetylglucosamine-1-phosphotransferase, gamma subunit |
Gene Aliases: |
6430527N14Rik, A830081F19, AU067667, AU067744, Mdcp1, Tce7 |
Location: |
Chr.17:25234317-25240170 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnptg | NM_172529.3 | 11 | 1100 | NP_766117.2 |
| AK044021.1 | 10 | 1180 | ||
| AK214503.1 | 1 | 189 | ||
| AK218936.1 | 1 | 189 | ||
| BC055872.1 | 11 | 1100 | AAH55872.1 |
Target Gene Details
Entrez Gene ID: | 74154 |
Gene Name: | unkempt-like (Drosophila) |
Gene Aliases: |
1300004G08Rik |
Location: |
Chr.17:25188344-25234443 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unkl | NM_001197024.1 | NP_001183953.1 | ||
| NM_001290736.1 | NP_001277665.1 | |||
| NM_028789.3 | NP_083065.1 | |||
| AK004898.1 | BAB23653.1 | |||
| BC089378.1 | AAH89378.1 |
More Information
Set Membership: |
|

Genomic Map