Assay Details
Target Gene Details
Entrez Gene ID: | 68799 |
Gene Name: | repulsive guidance molecule family member B |
Gene Aliases: |
1110059F19Rik, DRAGON |
Location: |
Chr.17:15804661-15831329 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgmb | NM_178615.3 | 2 | 743 | NP_848730.2 |
| AJ557514.1 | 2 | 743 | CAD89719.1 | |
| AK004310.1 | 2 | 295 | ||
| AK030386.1 | 2 | 697 | BAC26937.1 | |
| AK035713.1 | 2 | 690 | BAC29163.1 | |
| AK046637.1 | 2 | 691 | ||
| AK047390.1 | 2 | 700 | ||
| AK080819.1 | 2 | 690 | BAC38034.1 | |
| BC030405.1 | 2 | 726 | ||
| BC096024.1 | 2 | 690 | ||
| BC138890.1 | 2 | 566 |

Genomic Map