Assay Details
Target Gene Details
Entrez Gene ID: | 171236 |
Gene Name: | vomeronasal 1 receptor 233 |
Gene Aliases: |
V1rf5 |
Location: |
Chr.17:20993727-20994686 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vmn1r233 | NM_134202.1 | 1 | 800 | NP_598963.1 |
| BC115528.1 | 1 | 796 |
More Information
Set Membership: |
|

Genomic Map