Assay Details
Target Gene Details
Entrez Gene ID: | 70425 |
Gene Name: | casein kinase 1, gamma 3 |
Gene Aliases: |
3300002K07Rik, C330049O21Rik |
Location: |
Chr.18:53862082-53955684 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csnk1g3 | NM_152809.2 | 13 | 3168 | NP_690022.2 |
| AK014371.2 | 1 | 1200 | ||
| AK032675.1 | 7 | 1723 | BAC27983.1 | |
| AK034842.1 | 3 | 1349 | BAC28850.1 | |
| BC033601.1 | 10 | 2183 |
More Information
Set Membership: |
|

Genomic Map