Assay Details
Target Gene Details
Entrez Gene ID: | 13026 |
Gene Name: | phosphate cytidylyltransferase 1, choline, alpha isoform |
Gene Aliases: |
CTalpha, Cctalpha, Ctpct, Cttalpha |
Location: |
Chr.16:32430921-32475065 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcyt1a | NM_001163159.1 | 9 | 3064 | NP_001156631.1 |
| NM_001163160.1 | 9 | 2998 | NP_001156632.1 | |
| NM_009981.4 | 9 | 2900 | NP_034111.1 | |
| AK044585.1 | 9 | 2717 | ||
| AK163685.1 | 1 | 1704 | ||
| AK168266.1 | 1 | 227 | BAE40214.1 |
More Information
Set Membership: |
|

Genomic Map