Assay Details
Target Gene Details
Entrez Gene ID: | 21380 |
Gene Name: | T-box 1 |
Gene Aliases: |
- |
Location: |
Chr.16:18581704-18590671 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbx1 | NM_001285472.1 | 3 | 365 | NP_001272401.1 |
| NM_001285476.1 | 1 | 293 | NP_001272405.1 | |
| NM_011532.2 | 1 | 293 | NP_035662.1 | |
| AF326960.2 | 1 | 293 | AAG61088.1 | |
| AF349658.1 | 3 | 365 | AAK14407.1 | |
| BC139474.1 | 1 | 293 |
More Information
Set Membership: |
|

Genomic Map