Assay Details
Target Gene Details
Entrez Gene ID: | 67470 |
Gene Name: | ATP-binding cassette, sub-family G (WHITE), member 8 |
Gene Aliases: |
1300003C16Rik, AI114946, sterolin-2 |
Location: |
Chr.17:84676302-84700333 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcg8 | NM_001286005.1 | 10 | 1397 | NP_001272934.1 |
| NM_026180.3 | 11 | 1649 | NP_080456.1 | |
| NR_104382.1 | 11 | 1646 | ||
| AF324495.1 | 11 | 1639 | AAK84079.1 | |
| AK004871.1 | 11 | 1606 | BAB23630.1 | |
| AK050938.1 | 10 | 1399 | ||
| AY196215.1 | 11 | 1636 | AAO45095.1 | |
| AY196216.1 | 11 | 1636 | AAO45096.1 | |
| BC138484.1 | 11 | 1613 | ||
| BC145293.1 | 11 | 1613 |
More Information
Set Membership: |
|

Genomic Map