Assay Details
Target Gene Details
Entrez Gene ID: | 22339 |
Gene Name: | vascular endothelial growth factor A |
Gene Aliases: |
Vegf, Vpf |
Location: |
Chr.17:46016993-46032377 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vegfa | NM_001025250.3 | NP_001020421.2 | ||
| NM_001025257.3 | NP_001020428.2 | |||
| NM_001110266.1 | 1 | 474 | NP_001103736.1 | |
| NM_001110267.1 | 1 | 474 | NP_001103737.1 | |
| NM_001110268.1 | 1 | 474 | NP_001103738.1 | |
| NM_001287056.1 | NP_001273985.1 | |||
| NM_001287057.1 | NP_001273986.1 | |||
| NM_001287058.1 | NP_001273987.1 | |||
| NM_009505.4 | NP_033531.3 | |||
| AY120866.1 | AAM55477.1 | |||
| AY707864.1 | AAU11325.1 | |||
| AY750956.1 | AAU95484.1 | |||
| AY750957.1 | AAU95485.1 | |||
| AY756068.1 | AAV32504.1 | |||
| BC061468.1 | AAH61468.1 | |||
| BQ554097.1 | 1 | 119 | ||
| BQ832724.1 | ||||
| BU920197.1 | 1 | 474 | ||
| EF028705.1 | ||||
| M95200.1 | AAA40547.1 | |||
| S37052.1 | AAB22252.1 | |||
| S38083.1 | AAB22253.1 | |||
| S38100.1 | AAB22254.1 |
More Information
Set Membership: |
|

Genomic Map