Assay Details
Target Gene Details
Entrez Gene ID: | 240239 |
Gene Name: | G protein-coupled receptor 151 |
Gene Aliases: |
C130082O03Rik, GalRL, PGR7, nGPCR-2037 |
Location: |
Chr.18:42578020-42579652 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr151 | NM_181543.1 | 1 | 1090 | NP_853521.1 |
| AJ564167.1 | 1 | 1090 | CAD91896.1 | |
| AK048591.1 | 1 | 748 | BAC33383.1 | |
| AY351677.1 | 1 | 1037 | AAQ62568.1 | |
| BC120520.1 | 1 | 1070 | ||
| BC120526.1 | 1 | 1070 |
More Information
Set Membership: |
|

Genomic Map