Assay Details
Target Gene Details
Entrez Gene ID: | 14815 |
Gene Name: | nuclear receptor subfamily 3, group C, member 1 |
Gene Aliases: |
GR, Grl-1, Grl1 |
Location: |
Chr.18:39410545-39519416 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr3c1 | NM_008173.3 | 1 | 621 | NP_032199.3 |
| AK037733.1 | 1 | 501 | ||
| AK155200.1 | 3 | 703 | BAE33113.1 | |
| AK156323.1 | 3 | 697 | BAE33674.1 | |
| BC016462.1 | 3 | 704 | ||
| BC129912.1 | 2 | 599 | ||
| BC129913.1 | 2 | 599 | ||
| CN720992.1 | 1 | 156 | ||
| DQ504162.1 | 1 | 608 | ||
| HM236293.1 | 2 | 581 | ||
| X04435.1 | 2 | 599 | CAA28031.1 | |
| X13358.1 | 2 | 581 | CAA31738.1 | |
| X13359.1 | 2 | 581 | CAA31739.1 |

Genomic Map