Assay Details
Target Gene Details
Entrez Gene ID: | 14815 |
Gene Name: | nuclear receptor subfamily 3, group C, member 1 |
Gene Aliases: |
GR, Grl-1, Grl1 |
Location: |
Chr.18:39410545-39519416 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr3c1 | NM_008173.3 | 8 | 4287 | NP_032199.3 |
| AA124418.1 | 1 | 46 | ||
| AK037733.1 | 8 | 4168 | ||
| AK155200.1 | 10 | 4369 | BAE33113.1 | |
| AK156323.1 | 10 | 4363 | BAE33674.1 |

Genomic Map