Assay Details
Target Gene Details
Entrez Gene ID: | 67199 |
Gene Name: | prefoldin 1 |
Gene Aliases: |
2700086I23Rik, AA408327, AU044714 |
Location: |
Chr.18:36403679-36454495 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pfdn1 | NM_026027.3 | 4 | 684 | NP_080303.1 |
| AK002575.1 | BAB22200.1 | |||
| AK003330.1 | BAB22718.1 | |||
| AK010532.1 | BAB27010.1 | |||
| AK012575.1 | 4 | 685 | BAB28328.1 | |
| BC024693.1 | AAH24693.1 | |||
| BC094627.1 | AAH94627.1 |
More Information
Set Membership: |
|

Genomic Map