Assay Details
Target Gene Details
Entrez Gene ID: | 69481 |
Gene Name: | actin-like 9 |
Gene Aliases: |
1700029I08Rik, AV259599 |
Location: |
Chr.17:33432899-33434268 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actl9 | NM_183282.2 | 1 | 229 | NP_899105.2 |
| AK006498.1 | 1 | 233 | BAC25145.1 | |
| BU583640.1 | ||||
| BY083809.1 | 1 | 233 | ||
| CN841548.1 |
More Information
Set Membership: |
|

Genomic Map