Assay Details
Target Gene Details
Entrez Gene ID: | 53895 |
Gene Name: | caseinolytic mitochondrial matrix peptidase proteolytic subunit |
Gene Aliases: |
AU019820, D17Wsu160e |
Location: |
Chr.17:56990264-56996371 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clpp | NM_017393.2 | 6 | 821 | NP_059089.1 |
| AJ005253.1 | 6 | 789 | CAA06443.1 | |
| AK002238.1 | 6 | 760 | BAC24984.1 | |
| AK004024.1 | 6 | 770 | BAB23132.1 | |
| AK145765.1 | 6 | 783 | BAE26636.1 | |
| AK147719.1 | 6 | 749 | BAE28093.1 | |
| AK168053.1 | 6 | 768 | BAE40033.1 | |
| AK179220.1 | ||||
| AK186638.1 | ||||
| AK207558.1 | ||||
| AK216056.1 | ||||
| BC001998.1 | 6 | 780 | AAH01998.1 |
More Information
Set Membership: |
|

Genomic Map