Assay Details
Target Gene Details
Entrez Gene ID: | 21678 |
Gene Name: | TEA domain family member 3 |
Gene Aliases: |
DTEF-1, ETFR-1, TEAD-3, TEF-5, Tcf13r2 |
Location: |
Chr.17:28331671-28350805 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tead3 | NM_001098226.3 | 13 | 1867 | NP_001091696.2 |
| NM_001204156.1 | 13 | 2190 | NP_001191085.1 | |
| NM_011566.4 | 13 | 2031 | NP_035696.3 | |
| AF002670.1 | 13 | 1986 | AAC12263.1 | |
| AK087649.1 | 13 | 1869 | ||
| AK143551.1 | 12 | 2903 | ||
| BC021364.1 | 13 | 1911 | AAH21364.1 | |
| BC027383.1 | 4 | 942 | AAH27383.1 | |
| BC039984.1 | 4 | 977 | AAH39984.1 | |
| CB182196.1 | 2 | 736 | ||
| D87963.1 | 12 | 1701 | BAA13516.1 | |
| Y10027.1 | 12 | 1793 | CAA71136.1 |
More Information
Set Membership: |
|

Genomic Map