Assay Details
Target Gene Details
Entrez Gene ID: | 22339 |
Gene Name: | vascular endothelial growth factor A |
Gene Aliases: |
Vegf, Vpf |
Location: |
Chr.17:46016993-46032377 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vegfa | NM_001025250.3 | 1 | 364 | NP_001020421.2 |
| NM_001025257.3 | 1 | 364 | NP_001020428.2 | |
| NM_001287056.1 | 1 | 364 | NP_001273985.1 | |
| NM_001287057.1 | 1 | 364 | NP_001273986.1 | |
| NM_001287058.1 | 1 | 364 | NP_001273987.1 | |
| NM_009505.4 | 1 | 364 | NP_033531.3 | |
| AK131850.1 | 1 | 1086 | ||
| BY001490.1 | 1 | 365 | ||
| CK626177.1 | 1 | 349 |
More Information
Set Membership: |
|

Genomic Map