Assay Details
Target Gene Details
Entrez Gene ID: | 67365 |
Gene Name: | haloacid dehalogenase-like hydrolase domain containing 1A |
Gene Aliases: |
1700121L12Rik, GS1, Hdhd1 |
Location: |
Chr.18:50567656-50568699 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hdhd1a | NM_026108.3 | 1 | 697 | NP_080384.2 |
| AK007231.1 | 1 | 723 | BAB24906.1 | |
| AK014922.1 | 1 | 674 | BAB29622.1 | |
| BC048447.1 | 1 | 686 | AAH48447.1 |
More Information
Set Membership: |
|

Genomic Map