Assay Details
Target Gene Details
Entrez Gene ID: | 74131 |
Gene Name: | SAM and SH3 domain containing 3 |
Gene Aliases: |
1200013B08Rik, AW413946, SLY1 |
Location: |
Chr.X:48146451-48161565 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sash3 | NM_028773.3 | 8 | 2077 | NP_083049.1 |
| AK004734.2 | 8 | 2078 | BAB23516.1 | |
| AK156202.1 | 8 | 2142 | BAE33623.1 | |
| AK171017.1 | 8 | 2075 | BAE42187.1 | |
| BC028773.1 | 5 | 1639 | AAH28773.1 |

Genomic Map