Assay Details
Target Gene Details
Entrez Gene ID: | 56526 |
Gene Name: | septin 6 |
Gene Aliases: |
2810035H17Rik, C920001C06Rik, Sep6, mKIAA0128 |
Location: |
Chr.X:36911272-36989695 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sept6 | NM_001177323.2 | 10 | 2405 | NP_001170794.1 |
| NM_019942.5 | 10 | 2408 | NP_064326.2 | |
| AK048455.1 | 10 | 2230 | BAC33342.1 | |
| AK088614.1 | 10 | 2171 | BAC40453.1 | |
| AK160884.1 | 10 | 2202 | BAE36069.1 |
More Information
Set Membership: |
|

Genomic Map