Assay Details
Target Gene Details
Entrez Gene ID: | 22139 |
Gene Name: | transthyretin |
Gene Aliases: |
AA408768, AI787086, D17860, prealbumin |
Location: |
Chr.18:20665250-20674326 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttr | NM_013697.5 | 4 | 988 | NP_038725.1 |
| AK014454.1 | 4 | 825 | BAB29362.1 | |
| AK080757.1 | 1 | 233 | ||
| CD742426.1 | 1 | 250 | ||
| D89076.1 | 4 | 825 | BAA13757.1 |
More Information
Set Membership: |
|

Genomic Map