Assay Details
Target Gene Details
Entrez Gene ID: | 67951 |
Gene Name: | tubulin, beta 6 class V |
Gene Aliases: |
2310057H16Rik, BB220206 |
Location: |
Chr.18:67390731-67402749 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb6 | NM_026473.2 | 4 | 1508 | NP_080749.2 |
| AK009958.1 | 4 | 1519 | ||
| AK015255.1 | 1 | 774 | BAB29767.1 | |
| AK144815.1 | 4 | 1523 | BAE26077.1 | |
| AK151643.1 | 4 | 1505 | BAE30573.1 | |
| AK195708.1 | 1 | 113 | ||
| BC008225.1 | 4 | 1508 | AAH08225.1 |
More Information
Set Membership: |
|

Genomic Map