Assay Details
Target Gene Details
Entrez Gene ID: | 269053 |
Gene Name: | G protein-coupled receptor 152 |
Gene Aliases: |
A930009H15Rik, Gm673 |
Location: |
Chr.19:4139799-4145741 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr152 | NM_206973.2 | 2 | 2810 | NP_996856.1 |
| AK044225.1 | ||||
| AK044366.1 | BAC31886.1 | |||
| BC099681.1 | 2 | 2810 | AAH99681.1 |
More Information
Set Membership: |
|

Genomic Map