Assay Details
Target Gene Details
Entrez Gene ID: | 18089 |
Gene Name: | NK2 homeobox 3 |
Gene Aliases: |
Nkx-2.3, Nkx2.3, tinman |
Location: |
Chr.19:43612325-43615892 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx2-3 | NM_008699.2 | 2 | 1846 | NP_032725.1 |
| AK044871.1 | 2 | 1779 | BAC32125.1 | |
| BC072614.1 | 2 | 1846 | AAH72614.1 |
More Information
Set Membership: |
|

Genomic Map