Assay Details
Target Gene Details
Entrez Gene ID: | 56389 |
Gene Name: | syntaxin 5A |
Gene Aliases: |
0610031F24Rik, D19Ertd627e, Stx5 |
Location: |
Chr.19:8741424-8755642 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stx5a | NM_001167799.1 | NP_001161271.1 | ||
| NM_019829.4 | 1 | 463 | NP_062803.4 | |
| AK002722.1 | ||||
| AK135445.1 | 1 | 461 | ||
| AK166087.1 | 1 | 466 | BAE38563.1 | |
| AK170893.1 | 1 | 756 | ||
| AK193630.1 | ||||
| BC004849.1 | AAH04849.1 | |||
| BC021883.1 | 1 | 446 | AAH21883.1 | |
| CK389576.1 | ||||
| CN539678.1 |
More Information
Set Membership: |
|

Genomic Map