Assay Details
Target Gene Details
Entrez Gene ID: | 56389 |
Gene Name: | syntaxin 5A |
Gene Aliases: |
0610031F24Rik, D19Ertd627e, Stx5 |
Location: |
Chr.19:8741424-8755642 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stx5a | NM_001167799.1 | 11 | 1325 | NP_001161271.1 |
| NM_019829.4 | 10 | 1624 | NP_062803.4 | |
| AK002722.1 | 11 | 1304 | ||
| AK166087.1 | 10 | 1627 | BAE38563.1 | |
| AK178504.1 | ||||
| AK180075.1 | ||||
| AK208558.1 | ||||
| AK210992.1 | ||||
| AK214856.1 | ||||
| BC004849.1 | 11 | 1258 | AAH04849.1 | |
| BC021883.1 | 10 | 1607 | AAH21883.1 | |
| BQ030996.1 | 1 | 505 | ||
| DV649852.1 | 5 | 588 |
More Information
Set Membership: |
|

Genomic Map