Assay Details
Target Gene Details
Entrez Gene ID: | 30052 |
Gene Name: | proprotein convertase subtilisin/kexin type 1 inhibitor |
Gene Aliases: |
AI848336, KEP, PEN, PEN19, PEN20, Pan3, SAAS, SAASCT, bLEN, lLEN, proSAAS |
Location: |
Chr.X:7919822-7924410 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcsk1n | NM_013892.3 | 2 | 604 | NP_038920.2 |
| AF181560.1 | 2 | 604 | AAF22641.1 | |
| AF293356.1 | 2 | 588 | AAG09048.1 | |
| AK002969.1 | 2 | 531 | ||
| AK076137.1 | 2 | 570 | ||
| BC012263.1 | 2 | 514 | AAH12263.1 |
More Information
Set Membership: |
|

Genomic Map