Assay Details
Target Gene Details
Entrez Gene ID: | 12000 |
Gene Name: | arginine vasopressin receptor 2 |
Gene Aliases: |
ADHR, DI1, DIR, ND1, V2R, VPV2R |
Location: |
Chr.X:73890591-73894428 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Avpr2 | NM_001276298.1 | 2 | 306 | NP_001263227.1 |
| NM_001276299.1 | NP_001263228.1 | |||
| NM_019404.2 | 2 | 309 | NP_062277.1 | |
| AJ310753.1 | 2 | 309 | CAC34589.1 | |
| AK052712.1 | 2 | 714 | ||
| BC104728.1 | 2 | 298 | AAI04729.1 | |
| BC104729.1 | 2 | 301 | AAI04730.1 | |
| BC104731.1 | 2 | 301 | AAI04732.1 | |
| BC105662.1 | 2 | 301 | ||
| BC107005.2 | AAI07006.1 |
More Information
Set Membership: |
|

Genomic Map